Gene Transcripts
- Gene Symbol
- FAM221A
- Entrez Gene ID
- 340277
- Gene Name
- family with sequence similarity 221 member A
- Gene Aliases
- C7orf46
- Chromosome Location
- Chr. 7: 23680114 - 23703249 on Build GRCh38
- UniGene ID
- Hs.592178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127364.2 |
NP_001120836.1
|
5, 6 |
819 |
|
NM_001127365.2 |
NP_001120837.1
|
4, 5 |
645 |
|
NM_001300932.1 |
NP_001287861.1
|
5, 6 |
753 |
|
NM_199136.4 |
NP_954587.2
|
6, 7 |
927 |
|
XM_011515369.2 |
XP_011513671.1
|
6, 7 |
922 |
|
XM_011515370.2 |
XP_011513672.1
|
6, 7 |
919 |
|
XM_011515371.2 |
XP_011513673.1
|
5, 6 |
811 |
|
XM_011515372.2 |
XP_011513674.1
|
6, 7 |
1067 |
|
XM_017012134.1 |
XP_016867623.1
|
5, 6 |
814 |
|
XM_017012135.1 |
XP_016867624.1
|
5, 6 |
959 |
|
GenBank mRNA |
AK127774.1 |
- |
7, 8 |
3539 |
|
AL832127.1 |
- |
7, 8 |
3596 |
|
BC026000.1 |
- |
4, 5 |
577 |
|
BC042034.1 |
- |
5, 6 |
800 |
|
BC063130.1 |
- |
6, 7 |
841 |
30382529 |
View Full Product Details