Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052941.4 | NP_443173.2 | 8 | 1492 | |
GenBank mRNA | AF288814.1 | - | 8 | 1376 | |
AK131094.1 | - | 8 | 1405 | ||
AK304245.1 | - | 4 | 404 | ||
AK312417.1 | - | 8 | 1484 | ||
AL832576.2 | - | 8 | 1422 | ||
BC070055.1 | - | 8 | 1429 | 30345441 |