Gene Transcripts
- Gene Symbol
- CLEC4D
- Entrez Gene ID
- 338339
- Gene Name
- C-type lectin domain family 4 member D
- Gene Aliases
- CD368, CLEC-6, CLEC6, CLECSF8, MCL, MPCL
- Chromosome Location
- Chr. 12: 8513492 - 8522366 on Build GRCh38
- UniGene ID
- Hs.351811
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080387.4 |
NP_525126.2
|
5, 6 |
690 |
|
XM_011520632.2 |
XP_011518934.1
|
6, 7 |
659 |
|
GenBank mRNA |
AF411850.1 |
- |
5, 6 |
560 |
|
AK314071.1 |
- |
5, 6 |
677 |
|
AY115592.1 |
- |
5, 6 |
681 |
|
AY297446.1 |
- |
5, 6 |
689 |
|
AY486482.1 |
- |
5, 6 |
514 |
|
AY486483.1 |
- |
5, 6 |
514 |
|
BC032313.1 |
- |
5, 6 |
690 |
5218660 |
BI909298.1 |
- |
5, 6 |
640 |
|
View Full Product Details