Gene Transcripts

Gene Symbol
MYO3B
Entrez Gene ID
140469
Gene Name
myosin IIIB
Gene Aliases
Chromosome Location
Chr. 2: 170178145 - 170655325 on Build GRCh38
UniGene ID
Hs.671900
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083615.3 NP_001077084.2 4 529
NM_138995.4 NP_620482.3 4 529
NR_045682.1 4 529
NR_045683.1 4 529
NR_045684.1 4 529
XM_006712299.3 XP_006712362.1 4 417
XM_011510654.2 XP_011508956.1 4 417
XM_011510655.2 XP_011508957.1 4 417
GenBank mRNA AF369908.1 - 4 529
AF391554.1 - 4 529
AF391555.1 - 4 529
AF391556.1 - 4 529
AF391557.1 - 4 529
AF391558.1 - 4 529
AK055778.1 - 4 417
BC040019.1 - 3 384
BC136620.1 - 4 466 9020610
BC144307.1 - 4 466 9052827
BX647895.1 - 3 340

View Full Product Details