Gene Transcripts
- Gene Symbol
- ZDHHC15
- Entrez Gene ID
- 158866
- Gene Name
- zinc finger DHHC-type containing 15
- Gene Aliases
- MRX91
- Chromosome Location
- Chr. X: 75368427 - 75523626 on Build GRCh38
- UniGene ID
- Hs.253211
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146256.1 |
NP_001139728.1
|
3, 4 |
838 |
|
NM_144969.2 |
NP_659406.1
|
4, 5 |
865 |
|
XM_006724624.3 |
XP_006724687.1
|
4, 5 |
402 |
|
XM_017029296.1 |
XP_016884785.1
|
4, 5 |
402 |
|
GenBank mRNA |
AK056374.1 |
- |
4, 5 |
403 |
|
AK122885.1 |
- |
3, 4 |
587 |
|
AK128587.1 |
- |
3, 4 |
373 |
|
AK303896.1 |
- |
4, 5 |
403 |
|
BC103980.1 |
- |
4, 5 |
403 |
40016292 |
BC103981.1 |
- |
4, 5 |
403 |
40016293 |
BC103982.1 |
- |
4, 5 |
403 |
40016294 |
View Full Product Details