Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145025.4 | NP_659462.1 | 12 | 1339 | |
XM_017010387.1 | XP_016865876.1 | 7 | 925 | ||
GenBank mRNA | AK055538.1 | - | 12 | 1316 | |
AL832162.1 | - | 12 | 1318 | ||
BC022031.2 | - | 12 | 1344 |