Gene Transcripts

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location
Chr. 6: 39335100 - 39725405 on Build GRCh38
UniGene ID
Hs.588202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289020.1 NP_001275949.1 9 1123
NM_001289021.1 NP_001275950.1 9 1123
NM_145027.4 NP_659464.3 9 1123
XM_005248904.4 XP_005248961.1 9 1123
XM_011514357.2 XP_011512659.1 9 1048
XM_011514358.2 XP_011512660.1 9 1048
XM_011514359.2 XP_011512661.1 9 1048
XM_011514361.2 XP_011512663.1 9 1123
XM_017010427.1 XP_016865916.1 8 1013
XM_017010428.1 XP_016865917.1 4 473
GenBank mRNA AK131471.1 - 9 1123
BC117307.1 - 9 1053 40125858
BC143444.1 - 9 1053 9051952
BC143445.1 - 9 1053 9051953
BX649045.1 - 6 703

View Full Product Details