Gene Transcripts

Gene Symbol
HUS1B
Entrez Gene ID
135458
Gene Name
HUS1 checkpoint clamp component B
Gene Aliases
Chromosome Location
Chr. 6: 655939 - 656964 on Build GRCh38
UniGene ID
Hs.669039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148959.3 NP_683762.2 1 757
GenBank mRNA AF508547.1 - 1 737
BC101697.1 - 1 757 8069203
BC101699.1 - 1 757 8069205

View Full Product Details