Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286354.1 | NP_001273283.1 | 6 | 1637 | |
NM_001286355.1 | NP_001273284.1 | 5 | 1949 | ||
NM_148963.3 | NP_683766.2 | 6 | 2162 | ||
XM_017010475.1 | XP_016865964.1 | 7 | 4095 | ||
GenBank mRNA | AF502962.1 | - | 6 | 2140 | |
AY435125.1 | - | 6 | 2151 | ||
AY435126.1 | - | 6 | 1626 | ||
AY435127.1 | - | 5 | 1938 |