Gene Transcripts

Gene Symbol
TTC1
Entrez Gene ID
7265
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
TPR1
Chromosome Location
Chr. 5: 160009100 - 160065545 on Build GRCh38
UniGene ID
Hs.519718
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282500.1 NP_001269429.1 2 299
NM_003314.2 NP_003305.1 2 302
GenBank mRNA AK222641.1 - 2 253
AK222984.1 - 2 247
AK315262.1 - 2 257
BC000942.3 - 1 207 3446109
U46570.1 - 2 229

View Full Product Details