Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175066.3 | NP_778236.2 | 3 | 625 | |
XM_011538256.1 | XP_011536558.1 | 3 | 625 | ||
GenBank mRNA | AB593157.1 | - | 3 | 645 | |
AK097078.1 | - | 3 | 625 | ||
AL079273.1 | - | 1 | 73 | ||
BC029279.1 | - | 3 | 605 | ||
BC040185.2 | - | 3 | 598 | 4801253 |