Gene Transcripts

Gene Symbol
TRAPPC6B
Entrez Gene ID
122553
Gene Name
trafficking protein particle complex 6B
Gene Aliases
TPC6
Chromosome Location
Chr. 14: 39147811 - 39170430 on Build GRCh38
UniGene ID
Hs.13303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079537.1 NP_001073005.1 2 430
NM_177452.3 NP_803235.1 2 430
XM_011536409.2 XP_011534711.1 3 450
XM_017020964.1 XP_016876453.1 2 403
GenBank mRNA AK056690.1 - 2 400
AK310730.1 - 2 207
AK315900.1 - 2 325
AL833179.1 - 2 314
BC047328.1 - 2 314 5534151
BX161510.1 - 2 322
BX248013.1 - 2 353

View Full Product Details