Gene Transcripts

Gene Symbol
SERINC5
Entrez Gene ID
256987
Gene Name
serine incorporator 5
Gene Aliases
C5orf12, TPO1
Chromosome Location
Chr. 5: 80111227 - 80256082 on Build GRCh38
UniGene ID
Hs.288232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174071.2 NP_001167542.1 11 1325
NM_001174072.2 NP_001167543.1 11 1325
NM_178276.6 NP_840060.1 11 1325
NR_126060.1 11 1319
NR_126061.1 11 1325
XM_011543304.1 XP_011541606.1 11 1290
XM_017009323.1 XP_016864812.1 11 1271
XM_017009324.1 XP_016864813.1 11 1290
XM_017009325.1 XP_016864814.1 9 926
XM_017009326.1 XP_016864815.1 9 926
GenBank mRNA AF498273.1 - 11 1295
AK297467.1 - 11 1322
BC101280.2 - 11 1257 40024444
BC101281.3 - 11 1262 40024445
BC101283.2 - 11 1262 40024448

View Full Product Details