Gene Transcripts

Gene Symbol
PLAC4
Entrez Gene ID
191585
Gene Name
placenta specific 4
Gene Aliases
C21orf115, PRED78
Chromosome Location
Chr. 21: 41175231 - 41185239 on Build GRCh38
UniGene ID
Hs.472492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182832.2 NP_878252.2 1 435
GenBank mRNA AK092431.1 - 1 435
BC093685.1 - 1 380 7939530
BC101615.1 - 1 380 8069121
BC101617.1 - 1 380 8069123

View Full Product Details