Gene Transcripts

Gene Symbol
VBP1
Entrez Gene ID
7411
Gene Name
VHL binding protein 1
Gene Aliases
HIBBJ46, PFD3, PFDN3, VBP-1
Chromosome Location
Chr. X: 155197007 - 155239841 on Build GRCh38
UniGene ID
Hs.436803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303543.1 NP_001290472.1 1, 2 299
NM_003372.6 NP_003363.1 1, 2 148
GenBank mRNA AK313420.1 - 1, 2 116
BC046094.1 - 1, 2 101 6058145
BT019604.1 - 1, 2 89
DA610631.1 - 1, 2 148
U96759.1 - 1, 2 296
Y17394.1 - 1, 2 101

View Full Product Details