Gene Transcripts

Gene Symbol
NAV1
Entrez Gene ID
89796
Gene Name
neuron navigator 1
Gene Aliases
POMFIL3, STEERIN1, UNC53H1
Chromosome Location
Chr. 1: 201648322 - 201826974 on Build GRCh38
UniGene ID
Hs.585374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167738.1 NP_001161210.1 2 265
NM_020443.4 NP_065176.3 4 1596
XM_006711609.2 XP_006711672.2 2 231
XM_006711610.2 XP_006711673.2 2 231
XM_006711611.1 XP_006711674.1 2 347
XM_011510097.1 XP_011508399.1 4 1275
XM_011510098.1 XP_011508400.1 4 1275
XM_011510099.1 XP_011508401.1 4 1275
XM_011510100.1 XP_011508402.1 4 1275
XM_011510101.1 XP_011508403.1 4 1275
XM_011510102.1 XP_011508404.1 4 1275
XM_017002751.1 XP_016858240.1 4 1275
XM_017002752.1 XP_016858241.1 2 231
XM_017002753.1 XP_016858242.1 4 1275
XM_017002754.1 XP_016858243.1 4 1275
GenBank mRNA AB032977.3 - 4 929
AJ488101.1 - 4 1127
AK056158.1 - 2 342
BX648460.1 - 2 232
DA431858.1 - 2 265

View Full Product Details