Gene Transcripts

Gene Symbol
TUBGCP6
Entrez Gene ID
85378
Gene Name
tubulin gamma complex associated protein 6
Gene Aliases
GCP-6, GCP6, MCCRP, MCCRP1, MCPHCR
Chromosome Location
Chr. 22: 50217689 - 50245024 on Build GRCh38
UniGene ID
Hs.336431
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020461.3 NP_065194.2 10 2434
GenBank mRNA AB051456.1 - 10 1828
AF272887.1 - 10 2027
AK000636.1 - 2 233
AL137665.1 - 3 251
AL832617.2 - 10 2455
BC144488.1 - 10 2026 9053015
BC144489.1 - 10 2026
BC150613.1 - 10 2026 9056983
BC151209.1 - 10 2416 40147276

View Full Product Details