Gene Transcripts

Gene Symbol
KCTD15
Entrez Gene ID
79047
Gene Name
potassium channel tetramerization domain containing 15
Gene Aliases
Chromosome Location
Chr. 19: 33795540 - 33815761 on Build GRCh38
UniGene ID
Hs.221873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024076.2 NP_076981.2 7 1569
GenBank mRNA BC001185.1 - 7 1553
BC009335.2 - 7 1553 4128019

View Full Product Details