Gene Transcripts

Gene Symbol
HSPBAP1
Entrez Gene ID
79663
Gene Name
HSPB1 associated protein 1
Gene Aliases
PASS1
Chromosome Location
Chr. 3: 122739997 - 122793829 on Build GRCh38
UniGene ID
Hs.29169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320728.1 NP_001307657.1 6 926
NM_024610.5 NP_078886.2 7 1002
XM_011513130.2 XP_011511432.1 8 920
XM_011513132.2 XP_011511434.2 7 761
XM_017007179.1 XP_016862668.1 6 826
XM_017007180.1 XP_016862669.1 3 464
GenBank mRNA AF400663.1 - 7 969
AK026276.1 - 1 701
AK289410.1 - 7 1002
BC011897.2 - 7 986 3949150
BC017763.1 - 1 50

View Full Product Details