Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024775.9 | NP_079051.9 | 3 | 558 | |
GenBank mRNA | AF453443.1 | - | 2 | 442 | |
AK027112.1 | - | 3 | 529 | ||
AK315627.1 | - | 3 | 537 | ||
BC018195.1 | - | 3 | 499 | 3863118 | |
BC029584.1 | - | 6 | 1075 | ||
BM471707.1 | - | 3 | 504 |