Gene Transcripts

Gene Symbol
TMEM254
Entrez Gene ID
80195
Gene Name
transmembrane protein 254
Gene Aliases
C10orf57, bA369J21.6
Chromosome Location
Chr. 10: 80078646 - 80092552 on Build GRCh38
UniGene ID
Hs.169982
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270367.1 NP_001257296.1 4 757
NM_001270368.1 NP_001257297.1 4 770
NM_001270369.1 NP_001257298.1 5 809
NM_001270370.1 NP_001257299.1 6 816
NM_001270371.1 NP_001257300.1 5 831
NM_001270372.1 NP_001257301.1 4 749
NM_001270373.1 NP_001257302.1 5 788
NM_001270374.1 NP_001257303.1 5 1138
NM_025125.3 NP_079401.2 4 736
NR_072984.1 5 753
NR_072985.1 5 760
NR_072986.1 4 658
NR_072987.1 5 710
XM_006717978.2 XP_006718041.1 5 1088
GenBank mRNA AK023325.1 - 4 712
AK222536.1 - 4 717
AK300490.1 - 4 767
AK309980.1 - 4 744
AK310160.1 - 5 718
AK311563.1 - 5 726
AL832397.1 - 4 633
AL832483.1 - 4 744
BC022252.1 - 4 712 4668550
BX537803.1 - 5 718

View Full Product Details