Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001319960.1 | NP_001306889.1 | 21 | 2927 | |
NM_025191.3 | NP_079467.3 | 20 | 2879 | ||
NR_135118.1 | 21 | 2946 | |||
XM_005245499.1 | XP_005245556.1 | 20 | 2863 | ||
XM_011510012.1 | XP_011508314.1 | 19 | 2802 | ||
XM_017002397.1 | XP_016857886.1 | 19 | 2799 | ||
GenBank mRNA | AF288393.1 | - | 20 | 2542 | |
AK315118.1 | - | 20 | 2823 | ||
AL117441.1 | - | 6 | 962 | ||
BC016464.1 | - | 1 | 63 | ||
BC144149.1 | - | 21 | 2587 | 9052669 | |
BC144150.1 | - | 21 | 2606 |