Gene Transcripts

Gene Symbol
PRR3
Entrez Gene ID
80742
Gene Name
proline rich 3
Gene Aliases
CAT56
Chromosome Location
Chr. 6: 30556709 - 30564696 on Build GRCh38
UniGene ID
Hs.118354
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077497.2 NP_001070965.1 3 1983
NM_025263.3 NP_079539.2 4 2046
GenBank mRNA AK074531.1 - 3 1412
AK131382.1 - 2 3374
U63336.1 - 4 1869

View Full Product Details