Gene Transcripts
- Gene Symbol
- VASP
- Entrez Gene ID
- 7408
- Gene Name
- vasodilator-stimulated phosphoprotein
- Gene Aliases
- Chromosome Location
- Chr. 19: 45507430 - 45526989 on Build GRCh38
- UniGene ID
- Hs.515469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003370.3 |
NP_003361.1
|
7, 8 |
1174 |
|
XM_005259199.1 |
XP_005259256.1
|
7, 8 |
1159 |
|
XM_005259200.1 |
XP_005259257.1
|
7, 8 |
1162 |
|
XM_017027200.1 |
XP_016882689.1
|
7, 8 |
1159 |
|
GenBank mRNA |
AK314812.1 |
- |
7, 8 |
1141 |
|
BC026019.1 |
- |
7, 8 |
1111 |
3345963 |
BC038224.1 |
- |
7, 8 |
1092 |
5210128 |
KU178381.1 |
- |
6, 7 |
832 |
|
KU178382.1 |
- |
7, 8 |
676 |
|
KU178383.1 |
- |
7, 8 |
673 |
|
KU178384.1 |
- |
6, 7 |
559 |
|
KU178385.1 |
- |
4, 5 |
381 |
|
Z46389.1 |
- |
7, 8 |
1086 |
|
View Full Product Details