Gene Transcripts

Gene Symbol
TRIM52
Entrez Gene ID
84851
Gene Name
tripartite motif containing 52
Gene Aliases
RNF102
Chromosome Location
Chr. 5: 181254077 - 181261160 on Build GRCh38
UniGene ID
Hs.458412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032765.2 NP_116154.1 2 1999
XM_005266000.3 XP_005266057.1 2 1406
XM_017009993.1 XP_016865482.1 2 1775
XM_017009994.1 XP_016865483.1 2 1601
GenBank mRNA AB209243.1 - 1 4522
AK026706.1 - 1 701
AK054802.1 - 2 1976
BC007372.2 - 2 1999 3636274
BC034154.1 - 1 1954

View Full Product Details