Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 2, 3 | 523 | |
XM_005268209.3 | XP_005268266.1 | 2, 3 | 327 | ||
XM_017021765.1 | XP_016877254.1 | 2, 3 | 326 | ||
GenBank mRNA | AB082526.1 | - | 2, 3 | 495 | |
AK128693.1 | - | 3, 4 | 459 | ||
AY048580.1 | - | 2, 3 | 389 | ||
AY080896.1 | - | 2, 3 | 401 | ||
BC093881.1 | - | 2, 3 | 470 | 7939726 | |
BC112101.1 | - | 2, 3 | 470 | 8327569 |