Gene Transcripts

Gene Symbol
RSPH1
Entrez Gene ID
89765
Gene Name
radial spoke head 1 homolog
Gene Aliases
CT79, RSP44, RSPH10A, TSA2, TSGA2
Chromosome Location
Chr. 21: 42472486 - 42496354 on Build GRCh38
UniGene ID
Hs.661069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080860.3 NP_543136.1 2 316
XM_011529786.1 XP_011528088.1 2 208
GenBank mRNA AB006536.1 - 2 316
AB041016.1 - 2 253
AK057315.1 - 2 208
AK314746.1 - 2 153
BC101519.1 - 2 195 8069025
BC113367.1 - 2 195 8322419

View Full Product Details