Gene Transcripts

Gene Symbol
ASB16
Entrez Gene ID
92591
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
Chromosome Location
Chr. 17: 44170706 - 44179083 on Build GRCh38
UniGene ID
Hs.534517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080863.4 NP_543139.4 2 591
GenBank mRNA AF403034.1 - 1 206
AK054727.1 - 2 591
AK314595.1 - 2 583
AY557346.1 - 2 507
BC075088.2 - 2 540 30915388
BC118584.1 - 2 573 40084995
BC118585.1 - 2 573 40084992

View Full Product Details