Gene Transcripts

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Chromosome Location
Chr. 9: 127451486 - 127503501 on Build GRCh38
UniGene ID
Hs.495188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005373.3 NP_001005373.1 11 1052
NM_001005374.3 NP_001005374.1 10 757
NM_001190723.2 NP_001177652.1 11 1052
NM_138361.5 NP_612370.3 10 1311
XM_006717316.3 XP_006717379.1 11 1052
XM_017015283.1 XP_016870772.1 11 2051
XM_017015284.1 XP_016870773.1 11 1052
GenBank mRNA AK056203.1 - 11 1034
AK091589.1 - 10 1059
AK226106.1 - 11 1038
AY358830.1 - 11 1017
BC009239.2 - 10 757 2988664

View Full Product Details