Gene Transcripts

Gene Symbol
EPS8L3
Entrez Gene ID
79574
Gene Name
EPS8 like 3
Gene Aliases
EPS8R3
Chromosome Location
Chr. 1: 109750080 - 109764022 on Build GRCh38
UniGene ID
Hs.485352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319952.1 NP_001306881.1 8 921
NM_024526.3 NP_078802.2 9 973
NM_133181.3 NP_573444.2 9 973
NM_139053.2 NP_620641.1 9 976
XM_011542132.1 XP_011540434.1 9 898
XM_011542133.1 XP_011540435.1 9 895
XM_011542134.2 XP_011540436.1 8 897
XM_011542135.2 XP_011540437.1 9 1015
XM_017002327.1 XP_016857816.1 9 898
XM_017002328.1 XP_016857817.1 9 898
XM_017002329.1 XP_016857818.1 9 874
GenBank mRNA AK025175.1 - 9 875
AK225235.1 - 9 875
AK290261.1 - 9 900
AK292198.1 - 9 897
AK307932.1 - 8 819
AK308196.1 - 8 715
AK310979.1 - 9 960
AY074930.1 - 9 859
BC012926.1 - 9 896 3853503

View Full Product Details