Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003356.3 | NP_003347.1 | 5 | 926 | |
NM_022803.2 | NP_073714.1 | 5 | 926 | ||
GenBank mRNA | AF001787.1 | - | 5 | 754 | |
AF011449.1 | - | 4 | 669 | ||
AK092685.1 | - | 5 | 573 | ||
AK290630.1 | - | 5 | 774 | ||
AK291889.1 | - | 5 | 740 | ||
AK309873.1 | - | 5 | 774 | ||
KU178372.1 | - | 4 | 571 | ||
U82818.1 | - | 5 | 724 | ||
U84763.1 | - | 5 | 724 |