Gene Transcripts

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
Chromosome Location
Chr. 8: 66112667 - 66175485 on Build GRCh38
UniGene ID
Hs.85524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033058.2 NP_149047.2 11 2619
NM_184085.1 NP_908973.1 10 2531
NM_184086.1 NP_908974.1 9 2243
NM_184087.1 NP_908975.1 5 1610
XM_005251316.4 XP_005251373.1 12 3553
XM_011517613.2 XP_011515915.1 12 3556
XM_011517614.2 XP_011515916.1 12 3479
XM_011517615.2 XP_011515917.1 13 3644
XM_011517616.2 XP_011515918.1 11 3268
XM_011517617.2 XP_011515919.1 10 2613
XM_017013908.1 XP_016869397.1 10 2249
XM_017013909.1 XP_016869398.1 9 2068
GenBank mRNA AK091310.1 - 9 2393

View Full Product Details