Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017925.6 | NP_060395.5 | 10 | 1481 | |
NR_073201.3 | 10 | 1481 | |||
XM_006716804.3 | XP_006716867.1 | 10 | 1449 | ||
XM_006716805.3 | XP_006716868.1 | 10 | 1449 | ||
XM_011517959.2 | XP_011516261.1 | 10 | 1449 | ||
XM_017014917.1 | XP_016870406.1 | 10 | 1449 | ||
XM_017014918.1 | XP_016870407.1 | 11 | 1649 | ||
XM_017014919.1 | XP_016870408.1 | 11 | 1616 | ||
GenBank mRNA | AK125842.1 | - | 4 | 343 | |
BC144484.1 | - | 6 | 713 | ||
BC144485.1 | - | 6 | 713 |