Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018229.3 | NP_060699.3 | 8 | 2085 | |
NR_026895.1 | 7 | 1439 | |||
XM_006720196.2 | XP_006720259.1 | 8 | 1714 | ||
XM_011536940.2 | XP_011535242.1 | 9 | 2057 | ||
GenBank mRNA | AK001675.1 | - | 8 | 1930 | |
AK172824.1 | - | 7 | 1148 | ||
AL136685.1 | - | 8 | 2064 |