Gene Transcripts

Gene Symbol
WDR12
Entrez Gene ID
55759
Gene Name
WD repeat domain 12
Gene Aliases
YTM1
Chromosome Location
Chr. 2: 202876958 - 202912226 on Build GRCh38
UniGene ID
Hs.73291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018256.3 NP_060726.3 11 1862
XM_011511469.2 XP_011509771.1 10 1739
GenBank mRNA AF242546.1 - 11 1245
AK001743.1 - 11 1299
AK022781.1 - 11 1307
AK022782.1 - 11 1307
AK056092.1 - 11 1862
AK299489.1 - 8 832
BC008082.2 - 11 1242 3050219
EF011617.1 - 11 1241

View Full Product Details