Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 1420 | |
NM_001145266.1 | NP_001138738.1 | 3 | 1323 | ||
NM_018389.4 | NP_060859.4 | 2 | 1694 | ||
XM_011520202.2 | XP_011518504.1 | 2 | 581 | ||
GenBank mRNA | AF323970.1 | - | 2 | 982 | |
AF326199.1 | - | 2 | 1591 | ||
AK000527.1 | - | 1 | 239 | ||
AK002182.1 | - | 3 | 1096 | ||
AK027394.1 | - | 2 | 1671 | ||
AK074929.1 | - | 2 | 1695 | ||
AK091245.1 | - | 2 | 2378 | ||
AK225428.1 | - | 3 | 1118 | ||
AK315473.1 | - | 2 | 1396 | ||
BC001427.2 | - | 2 | 999 | 3139599 |