Gene Transcripts

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
Chromosome Location
Chr. 14: 88385638 - 88470350 on Build GRCh38
UniGene ID
Hs.525518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040428.3 NP_001035518.1 6, 7 999
NM_018418.4 NP_060888.2 7, 8 1095
XM_017021452.1 XP_016876941.1 7, 8 1260
XM_017021454.1 XP_016876943.1 8, 9 1128
XM_017021455.1 XP_016876944.1 7, 8 1061
XM_017021456.1 XP_016876945.1 8, 9 1074
XM_017021457.1 XP_016876946.1 6, 7 1199
GenBank mRNA AF144487.3 - 7, 8 1075
AK055864.1 - 7 2667
AK290631.1 - 6, 7 970
AL136604.1 - 6, 7 979
BC008656.1 - 2, 3 527
BC090875.1 - 7, 8 1382 30383306
CR533563.1 - 6, 7 824
CR936777.1 - 7, 8 1041
EU668353.1 - 7, 8 1071

View Full Product Details