Gene Transcripts

Gene Symbol
TMEM242
Entrez Gene ID
729515
Gene Name
transmembrane protein 242
Gene Aliases
BM033, C6orf35
Chromosome Location
Chr. 6: 157289022 - 157324259 on Build GRCh38
UniGene ID
Hs.157212
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018452.5 NP_060922.2 4 2635
GenBank mRNA BC029130.1 - 4 1897

View Full Product Details