Gene Transcripts

Gene Symbol
SASH3
Entrez Gene ID
54440
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
753P9, CXorf9, HACS2, SH3D6C, SLY
Chromosome Location
Chr. X: 129779916 - 129795201 on Build GRCh38
UniGene ID
Hs.61469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018990.3 NP_061863.1 8 2361
XM_006724763.1 XP_006724826.1 7 2478
GenBank mRNA BC051881.1 - 8 2293

View Full Product Details