Gene Transcripts

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
Chromosome Location
Chr. 11: 124884218 - 124897935 on Build GRCh38
UniGene ID
Hs.524121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301088.1 NP_001288017.1 18 3172
NM_019055.5 NP_061928.4 18 3378
XM_006718861.1 XP_006718924.1 18 3197
XM_011542875.1 XP_011541177.1 12 2140
GenBank mRNA AF361473.1 - 18 3307
AK025195.1 - 6 1228
AK027852.1 - 18 3276
AK074163.1 - 15 4236
AK297845.1 - 12 1958
AY358083.1 - 18 3229
BC014995.1 - 17 2990
BC025675.1 - 5 2701
BC039602.1 - 18 3266 5590503
BC064643.1 - 8 1525
BC111562.1 - 18 3272 40068748
BC111748.1 - 18 3272 40068752

View Full Product Details