Gene Transcripts

Gene Symbol
SLC25A11
Entrez Gene ID
8402
Gene Name
solute carrier family 25 member 11
Gene Aliases
OGC, SLC20A4
Chromosome Location
Chr. 17: 4937130 - 4940167 on Build GRCh38
UniGene ID
Hs.184877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165417.1 NP_001158889.1 3 598
NM_001165418.1 NP_001158890.1 2 478
NM_003562.4 NP_003553.2 3 631
GenBank mRNA AF070548.1 - 3 485
AK290376.1 - 3 382
AK313010.1 - 3 487
BC006508.2 - 3 487 2960933
BC006519.2 - 3 487 2960933
BC016294.2 - 3 406 3911858
BC017170.2 - 3 487 2960933
BI752999.1 - 2 253
BM047391.1 - 3 346
CR456834.1 - 3 374

View Full Product Details