Gene Transcripts

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
Chromosome Location
Chr. 8: 86573608 - 86743675 on Build GRCh38
UniGene ID
Hs.154433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019098.4 NP_061971.3 10, 11 1227
XM_011517138.2 XP_011515440.1 8, 9 882
GenBank mRNA AF228520.1 - 7, 8 840
AF272900.1 - 10, 11 1225
BC150601.1 - 10, 11 1205 9056971

View Full Product Details