Gene Transcripts
- Gene Symbol
- LZTFL1
- Entrez Gene ID
- 54585
- Gene Name
- leucine zipper transcription factor like 1
- Gene Aliases
- BBS17
- Chromosome Location
- Chr. 3: 45823316 - 45916042 on Build GRCh38
- UniGene ID
- Hs.30824
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276378.1 |
NP_001263307.1
|
8, 9 |
965 |
|
NM_001276379.1 |
NP_001263308.1
|
7, 8 |
840 |
|
NM_020347.3 |
NP_065080.1
|
6, 7 |
684 |
|
NR_075080.1 |
|
5, 6 |
559 |
|
XM_006713207.2 |
XP_006713270.2
|
6, 7 |
669 |
|
XM_011533838.2 |
XP_011532140.1
|
6, 7 |
608 |
|
XM_017006644.1 |
XP_016862133.1
|
9, 10 |
1322 |
|
XM_017006645.1 |
XP_016862134.1
|
8, 9 |
1197 |
|
GenBank mRNA |
AJ297351.1 |
- |
6, 7 |
641 |
|
AK093705.1 |
- |
8, 9 |
1283 |
|
AK303416.1 |
- |
9, 10 |
736 |
|
BC025988.2 |
- |
6, 7 |
613 |
4838035 |
BC042483.1 |
- |
6, 7 |
644 |
4831113 |
BX640604.1 |
- |
6, 7 |
669 |
|
CN429018.1 |
- |
5, 6 |
469 |
|
DA408809.1 |
- |
5, 6 |
530 |
|
View Full Product Details