Gene Transcripts
- Gene Symbol
- ITGA10
- Entrez Gene ID
- 8515
- Gene Name
- integrin subunit alpha 10
- Gene Aliases
- PRO827
- Chromosome Location
- Chr. 1: 145891208 - 145910189 on Build GRCh38
- UniGene ID
- Hs.158237
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003637.4 |
NP_003628.2
|
3, 4 |
447 |
|
XM_017002622.1 |
XP_016858111.1
|
1, 2 |
404 |
|
XM_017002623.1 |
XP_016858112.1
|
1, 2 |
404 |
|
XM_017002624.1 |
XP_016858113.1
|
1, 2 |
404 |
|
XM_017002625.1 |
XP_016858114.1
|
1, 2 |
404 |
|
XM_017002626.1 |
XP_016858115.1
|
1, 2 |
404 |
|
XM_017002627.1 |
XP_016858116.1
|
1, 2 |
404 |
|
GenBank mRNA |
AF074015.1 |
- |
3, 4 |
293 |
|
AF112345.1 |
- |
3, 4 |
348 |
|
AK304183.1 |
- |
|
277 |
|
AK314255.1 |
- |
3, 4 |
296 |
|
AY358325.1 |
- |
3, 4 |
405 |
|
BM311010.1 |
- |
3, 4 |
450 |
|
View Full Product Details