Gene Transcripts
- Gene Symbol
- MS4A7
- Entrez Gene ID
- 58475
- Gene Name
- membrane spanning 4-domains A7
- Gene Aliases
- 4SPAN2, CD20L4, CFFM4, MS4A8
- Chromosome Location
- Chr. 11: 60378485 - 60395954 on Build GRCh38
- UniGene ID
- Hs.530735
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021201.4 |
NP_067024.1
|
5, 6 |
731 |
|
NM_206938.1 |
NP_996821.1
|
4, 5 |
596 |
|
NM_206939.1 |
NP_996822.1
|
5, 6 |
683 |
|
NM_206940.1 |
NP_996823.1
|
4, 5 |
548 |
|
GenBank mRNA |
AB026043.1 |
- |
5, 6 |
671 |
|
AF201951.1 |
- |
5, 6 |
642 |
|
AF237916.1 |
- |
5, 6 |
655 |
|
AF309653.1 |
- |
5, 6 |
684 |
|
AF350501.1 |
- |
5, 6 |
642 |
|
AK075106.1 |
- |
5, 6 |
735 |
|
AK297027.1 |
- |
4, 5 |
532 |
|
BC020673.1 |
- |
5, 6 |
655 |
4720044 |
CR457187.1 |
- |
4, 5 |
538 |
|
View Full Product Details