Gene Transcripts
- Gene Symbol
- ADAM12
- Entrez Gene ID
- 8038
- Gene Name
- ADAM metallopeptidase domain 12
- Gene Aliases
- ADAM12-OT1, CAR10, MCMP, MCMPMltna, MLTN, MLTNA
- Chromosome Location
- Chr. 10: 126012385 - 126388558 on Build GRCh38
- UniGene ID
- Hs.594351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288973.1 |
NP_001275902.1
|
3, 4 |
683 |
|
NM_001288974.1 |
NP_001275903.1
|
3, 4 |
683 |
|
NM_001288975.1 |
NP_001275904.1
|
3, 4 |
683 |
|
NM_003474.5 |
NP_003465.3
|
3, 4 |
683 |
|
NM_021641.4 |
NP_067673.2
|
3, 4 |
683 |
|
XM_017016705.1 |
XP_016872194.1
|
2, 3 |
595 |
|
GenBank mRNA |
AF023476.2 |
- |
3, 4 |
576 |
|
AF023477.2 |
- |
3, 4 |
576 |
|
AK127856.1 |
- |
3, 4 |
588 |
|
AK291629.1 |
- |
3, 4 |
502 |
|
AY358878.1 |
- |
3, 4 |
580 |
|
BC060804.1 |
- |
3, 4 |
622 |
30348514 |
DR004947.1 |
- |
3, 4 |
579 |
|
KF444157.1 |
- |
3, 4 |
683 |
|
View Full Product Details