Gene Transcripts

Gene Symbol
NGB
Entrez Gene ID
58157
Gene Name
neuroglobin
Gene Aliases
Chromosome Location
Chr. 14: 77265491 - 77271312 on Build GRCh38
UniGene ID
Hs.274363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021257.3 NP_067080.1 4 1807
GenBank mRNA AF422797.1 - 4 1807
AK098350.1 - 4 1409
BC032509.1 - 4 1514 5247233

View Full Product Details