Gene Transcripts

Gene Symbol
SLC17A9
Entrez Gene ID
63910
Gene Name
solute carrier family 17 member 9
Gene Aliases
C20orf59, POROK8, VNUT
Chromosome Location
Chr. 20: 62952647 - 62968597 on Build GRCh38
UniGene ID
Hs.512686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302643.1 NP_001289572.1 9 1091
NM_022082.3 NP_071365.3 8 1029
XM_011528978.2 XP_011527280.1 8 941
GenBank mRNA AK027065.1 - 1 18
AK074107.1 - 7 1825
AK095473.1 - 8 880
AK125597.1 - 8 1552
AK298503.1 - 8 935
BC025312.1 - 9 1031 4563089
BC027447.1 - 4 250
BC038593.1 - 8 871

View Full Product Details