Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144772.1 | NP_001138244.1 | 3 | 322 | |
NM_003580.3 | NP_003571.2 | 3 | 382 | ||
GenBank mRNA | AK292679.1 | - | 3 | 262 | |
AK294009.1 | - | 3 | 322 | ||
BC041124.1 | - | 3 | 364 | 5785460 | |
DR005814.1 | - | 3 | 388 | ||
X96586.1 | - | 3 | 180 |