Gene Transcripts

Gene Symbol
OSTF1
Entrez Gene ID
26578
Gene Name
osteoclast stimulating factor 1
Gene Aliases
OSF, SH3P2, bA235O14.1
Chromosome Location
Chr. 9: 75088480 - 75147201 on Build GRCh38
UniGene ID
Hs.494192
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012383.4 NP_036515.4 4 389
XM_006717053.3 XP_006717116.1 4 398
XM_006717055.3 XP_006717118.1 3 341
XM_011518524.2 XP_011516826.1 5 208
XM_011518525.2 XP_011516827.1 4 400
XM_017014622.1 XP_016870111.1 4 390
GenBank mRNA AK222596.1 - 4 333
AK291511.1 - 4 357
BC007459.1 - 4 295 4052054
BP312980.1 - 4 389
U63717.1 - 4 327

View Full Product Details